Learn More
BINDER™ Reinforced Inner Chamber
SDP
Catalog No. 08111766 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
08-111-766 Stainless Steel
1 options
Catalog No. 08-111-766 Supplier BINDER™ Supplier No. 80121732
Add to Cart
Add to Cart
For Use With: BF 400
  • Max. total load of 85 to 350 kg
Specifications
Description Inner chamber, reinforced - including 2 reinforced racks
For Use With (Equipment) BINDER Chambers BF 400
Material Stainless Steel
Type Reinforced Chamber
Includes 2 reinforced racks