Learn More
BINDER™ Reinforced Inner Chamber
SDP
Catalog No. 08111769 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
08-111-769 Stainless Steel
1 options
Catalog No. 08-111-769 Supplier BINDER™ Supplier No. 80121829
Add to Cart
Add to Cart
For Use With: BF 720
  • Max. total load of 85 to 350 kg
Specifications
Description Inner chamber, reinforced - max. total load 350 kg
For Use With (Equipment) BINDER Chambers BF 720
Holds Max. load 350 kg
Material Stainless Steel
Type Reinforced Chamber