Learn More
BINDER™ Reinforced Inner Chamber
SDP
Catalog No. p-200104358 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 1060
KB 130
KB 260
KB 470
KB 65
KB 720
KB PRO 1060
KB PRO 130
KB PRO 260
KB PRO 470
KB PRO 720
KBF 1060
KBF 130
KBF 260
KBF 470
KBF 720
KBF PRO 1060
KBF PRO 130
KBF PRO 260
KBF PRO 470
KBF PRO 720
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5737 KB 65
50-287-5590 KB 130
50-287-5620 KB 260
50-287-5654 KB 470
50-287-5611 KB 720
50-287-5693 KB 1060
50-287-5763 KB PRO 130
50-287-5747 KB PRO 260
50-287-5674 KB PRO 470
50-287-5633 KB PRO 720
50-287-5725 KB PRO 1060
50-287-5612 KBF 130
50-287-5764 KBF 260
50-287-5748 KBF 470
50-287-5659 KBF 720
50-287-5621 KBF 1060
50-287-5716 KBF PRO 130
50-287-5591 KBF PRO 260
50-287-5672 KBF PRO 470
50-287-5687 KBF PRO 720
50-287-5608 KBF PRO 1060
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 50-287-5737 Supplier BINDER™ Supplier No. 80122505
Only null left
Add to Cart
Add to Cart
Reinforced inner chamber for climate chambers and cooling incubation chambers.
  • Reinforced inner chamber for bulky and heavy samples.
  • Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
For Use With (Equipment) KB 65
Material Stainless Steel
Type Chamber Rack
Load Bearing Capacity (Metric) 100 kg