Learn More
Electron Microscopy Sciences Rubis Reinforced Plastic Tweezers Style K7, 120 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7819212
Encompass_Preferred
Made of PBTP (Polybutylenterephthalate) with 20% fiber-glass, the tweezers are resistant to chemicals and acids, highly isolating, and resists temperatures up to 200 degree C.
Tips: 120 mm
OAL: Pointed, flat-contact-tips
Catalog No. 50-238-07