Learn More
Electron Microscopy Sciences Rubis Reinforced Plastic Tweezers Style KR, 120 mm OAL
SDP
Catalog No. 5023809
Encompass_Preferred
Made of PBTP (Polybutylenterephthalate) with 20% fiber-glass, the tweezers are resistant to chemicals and acids, highly isolating, and resists temperatures up to 200 degree C.
Tips: 120 mm
OAL: Flat points, flat-contact-tips
Catalog No. 50-238-09 Supplier Electron Microscopy Sciences Supplier No. 7819401
Made of PBTP (Polybutylenterephthalate) with 20% fiber-glass, the tweezers are resistant to chemicals and acids, highly isolating, and resists temperatures up to 200 degree C.
Tips: 120 mm
OAL: Flat points, flat-contact-tips