Learn More
BINDER™ Reinforced Rack
SDP
Catalog No. 502875733 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 720, KBF/KBF PRO 720
KB/KBF/KB PRO/KBF PRO 260/470
KBF 130, KBF PRO 130
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5733 KB/KB PRO 720, KBF/KBF PRO 720
50-287-5645 KBF 130, KBF PRO 130
50-287-5582 KB/KBF/KB PRO/KBF PRO 260/470
50-287-5583 KB/KB PRO 1060, KBF/KBF PRO 1060
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-287-5733 Supplier BINDER™ Supplier No. 80122414
Add to Cart
Add to Cart
Reinforced shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KB/KB PRO 720, KBF/KBF PRO 720
Material Stainless Steel
Type Rack