Learn More
BINDER™ Reinforced Rack
SDP
Catalog No. 08111870 Shop All BINDER Inc. Products
Catalog No. 08-111-870 Supplier BINDER™ Supplier No. 80120345
Add to Cart
Add to Cart
Material: Stainless Steel; For Use With: FP 240, M 240
  • Manufactured from stainless steel
  • With one set of securing elements (4 pieces)
  • Maximum load of 50 kg or 70 kg
Specifications
Description Rack, reinforced - stainless steel, with fasteners (1 set of 4)
For Use With (Equipment) BINDER Chambers FP 240, M 240
Holds Max. 70 kg
Material Stainless Steel
Type Rack
Includes 1 set of 4 fasteners