Learn More
BINDER™ Reinforced Rack
SDP
Catalog No. 08111871 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
08-111-871 Stainless Steel
1 options
Catalog No. 08-111-871 Supplier BINDER™ Supplier No. 80120346
Add to Cart
Add to Cart
Material: Stainless Steel; For Use With: FED 400, FP 400, M 400
  • Manufactured from stainless steel
  • With one set of securing elements (4 pieces)
  • Maximum load of 50 kg or 70 kg
Specifications
Description Rack, reinforced - stainless steel, with fasteners (1 set of 4)
For Use With (Equipment) BINDER Chambers FED 400, FP 400, M 400
Holds Max. 70 kg
Material Stainless Steel
Type Rack
Includes 1 set of 4 fasteners