Learn More
Julabo USA Inc 1m reinforced rubber tubing, 12mm (1/2") ID
SDP
Catalog No. 507534147
Encompass_Preferred
Bath Accessories, Julabo, 1 m Reinforced tubing, 12 mm / 1/2″ inner dia. (-40 … +120°C). For use with FC1600, FC1600S, FC1600T, FC600, FC600S, FCW2500T, FCW600, FCW600S, FL1201, FL1701, FL601, FLW1701. (max 4.5bar). Priced per meter.
Catalog No. 50-753-4147 Supplier Julabo USA Inc Supplier No. 8930312
Bath Accessories, Julabo, 1 m Reinforced tubing, 12 mm / 1/2″ inner dia. (-40 … +120°C). For use with FC1600, FC1600S, FC1600T, FC600, FC600S, FCW2500T, FCW600, FCW600S, FL1201, FL1701, FL601, FLW1701. (max 4.5bar). Priced per meter.