Learn More
eMolecules​ rel-(1S,2S,5R)-8-Boc-2-methyl-3-oxo-8-aza-bicyclo[3.2.1]octane | 2716849-46-4 | | 1g
Supplier:  eMolecules​ 571063805
Encompass_Preferred
J&W PharmLab LLC | rel-(1S,2S,5R)-8-Boc-2-methyl-3-oxo-8-aza-bicyclo[3.2.1]octane | 1g | 571063805 | 60R1890 | 97.000 | 2716849-46-4 | | 239.315 | C13H21NO3
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-214-8252