Learn More
Medchemexpress LLC Rel-2-[[[(1-methylethyl)amino]carbonyl]amino]-N-[2-[[(1R,2S)-2-[[4-(methylthio)benzoyl]amino] | 445479-97-0 | 99.3% | 593.66 g/mol | C28H34F3N5O4S | 10 MG
SDP
Catalog No. 5000245236
Encompass_Preferred
A potent, selective small-molecule antagonist of the chemokine receptor CCR2 intended for research use. It displays high binding affinity (binding IC50 ≈ 5.1 nM) and functional antagonism in calcium flux and chemotaxis assays. The compound is supplied as a white to off-white solid with high purity and is readily soluble in DMSO for preparation of stock solutions.
  • High binding affinity to CCR2 (binding IC50 ≈ 5.1 nM).
  • Exhibits functional antagonism in calcium flux and chemotaxis assays (calcium flux IC50 ≈ 18 nM; chemotaxis IC50 ≈ 1 nM).
  • High purity as supplied.
  • Suitable for in vitro and in vivo pharmacology studies.
  • White to off-white solid, readily soluble in DMSO.
  • Stable under recommended frozen storage for long-term use.
Catalog No. 50-002-45236 Supplier Medchemexpress LLC Supplier No. HY10190810MG
A potent, selective small-molecule antagonist of the chemokine receptor CCR2 intended for research use. It displays high binding affinity (binding IC50 ≈ 5.1 nM) and functional antagonism in calcium flux and chemotaxis assays. The compound is supplied as a white to off-white solid with high purity and is readily soluble in DMSO for preparation of stock solutions.
  • High binding affinity to CCR2 (binding IC50 ≈ 5.1 nM).
  • Exhibits functional antagonism in calcium flux and chemotaxis assays (calcium flux IC50 ≈ 18 nM; chemotaxis IC50 ≈ 1 nM).
  • High purity as supplied.
  • Suitable for in vitro and in vivo pharmacology studies.
  • White to off-white solid, readily soluble in DMSO.
  • Stable under recommended frozen storage for long-term use.