Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rel-(3R,4S,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride | 1322625-07-9 | 98.0% | 243.69 | C8H18ClNO5 | 1 MG
SDP

Catalog No. 5000283871
Encompass_Preferred

rel-(3R,4S,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride is a piperidine-based triol salt reported as an impurity of the antidiabetic drug miglitol. Documented identifiers include CAS 1322625-07-9, formula C8H18ClNO5, and molecular weight 243.69 g/mol. The compound is supplied in small research pack sizes and characterized in the manufacturer datasheet and batch PDF.

  • Identified impurity of miglitol.
  • CAS 1322625-07-9.
  • Molecular formula C8H18ClNO5.
  • Molecular weight 243.69 g/mol.
  • Available in small research quantities (1 mg).
  • Reported purity 98.0% (supplier data).
  • Storage conditions per certificate of analysis.

Catalog No. 50-002-83871 Supplier Medchemexpress LLC Supplier No. HYW5859381MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rel-(3R,4S,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride is a piperidine-based triol salt reported as an impurity of the antidiabetic drug miglitol. Documented identifiers include CAS 1322625-07-9, formula C8H18ClNO5, and molecular weight 243.69 g/mol. The compound is supplied in small research pack sizes and characterized in the manufacturer datasheet and batch PDF.

  • Identified impurity of miglitol.
  • CAS 1322625-07-9.
  • Molecular formula C8H18ClNO5.
  • Molecular weight 243.69 g/mol.
  • Available in small research quantities (1 mg).
  • Reported purity 98.0% (supplier data).
  • Storage conditions per certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.