Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rel-(8R,8'R)-dimethyl-(7S,7'R)-bis(3,4-methylenedioxyphenyl)tetrahydro-furan | 178740-32-4 | 5 MG
SDP

Catalog No. 5000418997
Encompass_Preferred

rel-(8R,8'R)-Dimethyl-(7S,7'R)-bis(3,4-methylenedioxyphenyl)tetrahydro-furan is a chemical constituent found in the fruit of Myristica fragrans. Intended for research use only, this solid compound has a purity of 99.95% and is classified as a phenylpropanoid lignan.

  • Purity of 99.95%
  • Molecular weight of 340.37
  • Chemical formula: C₂₀H₂₀O₅
  • Appearance: Solid, white to off-white
  • Classified as a phenylpropanoid lignan
  • Derived from *Myristica fragrans* and *Schisandra chinensis (Turcz.) Baill.*
  • Recommended storage at 4°C, protected from light
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-18997 Supplier Medchemexpress LLC Supplier No. HYN22075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rel-(8R,8'R)-Dimethyl-(7S,7'R)-bis(3,4-methylenedioxyphenyl)tetrahydro-furan is a chemical constituent found in the fruit of Myristica fragrans. Intended for research use only, this solid compound has a purity of 99.95% and is classified as a phenylpropanoid lignan.

  • Purity of 99.95%
  • Molecular weight of 340.37
  • Chemical formula: C₂₀H₂₀O₅
  • Appearance: Solid, white to off-white
  • Classified as a phenylpropanoid lignan
  • Derived from *Myristica fragrans* and *Schisandra chinensis (Turcz.) Baill.*
  • Recommended storage at 4°C, protected from light
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.