Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rel-ACPT-I | 194918-76-8 | MFCD06795869 | 99.0% | 217.18 g·mol⁻1 | C8H11NO6 | 1 MG
SDP

Catalog No. 5000211451
Encompass_Preferred

rel-ACPT-I is a selective agonist of group III metabotropic glutamate receptors (mGluRs) with reported neuroprotective, anticonvulsant, and anxiolytic-like effects in preclinical studies. It is supplied as a high-purity research chemical for in vitro and in vivo pharmacology applications.

  • Selective agonist of group III metabotropic glutamate receptors.
  • Exhibits neuroprotective, anticonvulsant, and anxiolytic-like effects in preclinical models.
  • High purity (≥99.0%), suitable for pharmacological studies.
  • CAS number 194918-76-8 for unambiguous identification.
  • Molecular weight 217.18 g·mol⁻¹; formula C8H11NO6.
  • Soluble in water (~2 mg/mL) with sonication and warming.
  • Recommended storage: -20°C dry; solutions at -80°C (long term) or -20°C (short term).
  • Available in small research quantities, facilitating dose-response and in vivo dosing.

Catalog No. 50-002-11451 Supplier Medchemexpress LLC Supplier No. HY1013871MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rel-ACPT-I is a selective agonist of group III metabotropic glutamate receptors (mGluRs) with reported neuroprotective, anticonvulsant, and anxiolytic-like effects in preclinical studies. It is supplied as a high-purity research chemical for in vitro and in vivo pharmacology applications.

  • Selective agonist of group III metabotropic glutamate receptors.
  • Exhibits neuroprotective, anticonvulsant, and anxiolytic-like effects in preclinical models.
  • High purity (≥99.0%), suitable for pharmacological studies.
  • CAS number 194918-76-8 for unambiguous identification.
  • Molecular weight 217.18 g·mol⁻¹; formula C8H11NO6.
  • Soluble in water (~2 mg/mL) with sonication and warming.
  • Recommended storage: -20°C dry; solutions at -80°C (long term) or -20°C (short term).
  • Available in small research quantities, facilitating dose-response and in vivo dosing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.