Learn More
Aviva Systems Biology Corp REL Antibody - middle region (ARP38198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human REL. Alternate names: C-RelApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56559