Learn More
Aviva Systems Biology Corp Rel Antibody - middle region (ARP36966_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36966_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rel. It was validated on Western Blot. Rel,reticuloendotheliosis oncogene,c-Rel; Rel,proto-oncogene c-Rel,proto-oncogene c-Rel,OTTMUSP00000005467,C-Rel proto-oncogene proteinThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: c-RelApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-554