Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (rel)-AR234960 | 1408311-94-3 | 99.0% | 1 ML
SDP

Catalog No. 5000233277
Encompass_Preferred

A selective MAS receptor agonist for research use supplied as a 10 mM solution in DMSO (1 mL) or as solid samples. It activates ERK1/2 signaling and induces connective tissue growth factor expression.

  • Selective MAS receptor agonist.
  • Activates ERK1/2 signaling and downstream CTGF expression.
  • Supplied as 10 mM solution in DMSO (1 mL) or as multiple solid sizes.
  • High purity (≈99.0%).
  • Molecular weight 555.62, formula C27H30FN5O5S.
  • SDS available in multiple languages.
  • For laboratory research use only.

Catalog No. 50-002-33277 Supplier Medchemexpress LLC Supplier No. HY120006A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A selective MAS receptor agonist for research use supplied as a 10 mM solution in DMSO (1 mL) or as solid samples. It activates ERK1/2 signaling and induces connective tissue growth factor expression.

  • Selective MAS receptor agonist.
  • Activates ERK1/2 signaling and downstream CTGF expression.
  • Supplied as 10 mM solution in DMSO (1 mL) or as multiple solid sizes.
  • High purity (≈99.0%).
  • Molecular weight 555.62, formula C27H30FN5O5S.
  • SDS available in multiple languages.
  • For laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.