Learn More
Medchemexpress LLC (rel)-AR234960 | 1408311-94-3 | HY-120006A | 98.95% | 555.62 | 10 MG
SDP
Catalog No. 5000365145
Encompass_Preferred
(rel)-AR234960 is a selective and competitive agonist of the G protein-coupled receptor MAS. It activates the downstream ERK1/2 signaling pathway, leading to the expression of connective tissue growth factor (CTGF) and its downstream collagen subtype genes. This compound promotes collagen synthesis in cardiac fibroblasts via the MAS-ERK1/2-CTGF pathway and exacerbates extracellular matrix remodeling. It regulates the expression of cardiac fibrosis-related genes and can be utilized in heart failure research.
  • Selective and competitive MAS receptor agonist
  • Activates ERK1/2 signaling pathway
  • Induces CTGF and collagen gene expression
  • Promotes collagen synthesis in cardiac fibroblasts
  • Regulates cardiac fibrosis-related genes
  • Useful in heart failure research
Catalog No. 50-003-65145 Supplier Medchemexpress LLC Supplier No. 10MGHY120006A
(rel)-AR234960 is a selective and competitive agonist of the G protein-coupled receptor MAS. It activates the downstream ERK1/2 signaling pathway, leading to the expression of connective tissue growth factor (CTGF) and its downstream collagen subtype genes. This compound promotes collagen synthesis in cardiac fibroblasts via the MAS-ERK1/2-CTGF pathway and exacerbates extracellular matrix remodeling. It regulates the expression of cardiac fibrosis-related genes and can be utilized in heart failure research.
  • Selective and competitive MAS receptor agonist
  • Activates ERK1/2 signaling pathway
  • Induces CTGF and collagen gene expression
  • Promotes collagen synthesis in cardiac fibroblasts
  • Regulates cardiac fibrosis-related genes
  • Useful in heart failure research