Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rel-cis-pinic acid | 61774-58-1 | MFCD00136421 | 186.21 | C9H14O4 | 50 MG
SDP

Catalog No. 5000284488
Encompass_Preferred

Rel-cis-pinic acid is a research reagent and a primary oxidation product of α-pinene used in studies of atmospheric chemistry and biogenic emissions. It is a small organic acid (C9H14O4; MW 186.21 g/mol) typically supplied in milligram quantities for laboratory research.

  • Suitable for atmospheric and biogenic emission research.
  • Primary oxidation product of α-pinene.
  • Molecular formula C9H14O4.
  • Molecular weight 186.21 g/mol.
  • CAS number 61774-58-1.
  • Supplied in small research quantities; purity and storage details available in the certificate of analysis.

Catalog No. 50-002-84488 Supplier Medchemexpress LLC Supplier No. HYW58822050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rel-cis-pinic acid is a research reagent and a primary oxidation product of α-pinene used in studies of atmospheric chemistry and biogenic emissions. It is a small organic acid (C9H14O4; MW 186.21 g/mol) typically supplied in milligram quantities for laboratory research.

  • Suitable for atmospheric and biogenic emission research.
  • Primary oxidation product of α-pinene.
  • Molecular formula C9H14O4.
  • Molecular weight 186.21 g/mol.
  • CAS number 61774-58-1.
  • Supplied in small research quantities; purity and storage details available in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.