Learn More
Medchemexpress LLC (rel)-Lopinavir-d8 ((rel)-ABT-378-d8) | 1322625-54-6 | C37H40D8N4O5 | 1 MG
SDP
Catalog No. 5000356206
Encompass_Preferred
(rel)-Lopinavir-d8 ((rel)-ABT-378-d8) is the deuterium-labeled version of Lopinavir. Lopinavir is a highly potent, selective peptidomimetic inhibitor of the HIV-1 protease, with Kis of 1.3 to 3.6 pM for wild-type and mutant HIV protease. It acts by arresting the maturation of HIV-1, thus blocking its infectivity. Lopinavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 14.2 μM. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-56206 Supplier Medchemexpress LLC Supplier No. HY14588S1MG
(rel)-Lopinavir-d8 ((rel)-ABT-378-d8) is the deuterium-labeled version of Lopinavir. Lopinavir is a highly potent, selective peptidomimetic inhibitor of the HIV-1 protease, with Kis of 1.3 to 3.6 pM for wild-type and mutant HIV protease. It acts by arresting the maturation of HIV-1, thus blocking its infectivity. Lopinavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 14.2 μM. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.