Learn More
Medchemexpress LLC (rel)-ML-SI3 | 2108567-79-7 | 98.9% | 429.58 | 1 ML
SDP
Catalog No. 5000351689
Encompass_Preferred
(rel)-ML-SI3 is an active ingredient of ML-SI3, targeting three isoforms of TRPML. It functions as an inhibitor of TRPML1 and TRPML3, and a potent activator of TRPML2. This compound is supplied as a 10 mM solution in DMSO, ready for research applications.
  • Targets three isoforms of TRPML
  • Inhibitor of TRPML1 (IC50 = 3.1 μM) and TRPML3 (IC50 = 28.5 μM)
  • Potent activator of TRPML2 (EC50 = 3.3 μM)
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-51689 Supplier Medchemexpress LLC Supplier No. HY139426A10MM/1ML
(rel)-ML-SI3 is an active ingredient of ML-SI3, targeting three isoforms of TRPML. It functions as an inhibitor of TRPML1 and TRPML3, and a potent activator of TRPML2. This compound is supplied as a 10 mM solution in DMSO, ready for research applications.
  • Targets three isoforms of TRPML
  • Inhibitor of TRPML1 (IC50 = 3.1 μM) and TRPML3 (IC50 = 28.5 μM)
  • Potent activator of TRPML2 (EC50 = 3.3 μM)
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light