Learn More
Medchemexpress LLC (rel)-ML-SI3 | 2108567-79-7 | 98.9% | 429.58 | 25 MG
SDP
Catalog No. 5000349958
Encompass_Preferred
(rel)-ML-SI3 is one of the active ingredients of ML-SI3 (HY-139426), targeting three isoforms of TRPML. It functions as an inhibitor of TRPML1 and TRPML3, and a potent activator of TRPML2.
  • Targets three isoforms of TRPML
  • Inhibitor of TRPML1 and TRPML3
  • Potent activator of TRPML2
  • Supplied as a white to off-white solid
Catalog No. 50-003-49958 Supplier Medchemexpress LLC Supplier No. 25MGHY139426A
(rel)-ML-SI3 is one of the active ingredients of ML-SI3 (HY-139426), targeting three isoforms of TRPML. It functions as an inhibitor of TRPML1 and TRPML3, and a potent activator of TRPML2.
  • Targets three isoforms of TRPML
  • Inhibitor of TRPML1 and TRPML3
  • Potent activator of TRPML2
  • Supplied as a white to off-white solid