Learn More
Medchemexpress LLC Rel-N-((1R,2R)-2-(4-(2-methoxyphenyl)piperazin-1-yl)cyclohexyl)benzenesulfonamide | 2108567-79-7 | MFCD32690912 | 98.9% | 429.58 g·mol⁻¹ | C23H31N3O3S | 5 MG
SDP
Catalog No. 5000222186
Encompass_Preferred
(rel)-ML-SI3 is a small-molecule TRPML1/3 inhibitor intended for preclinical and in vitro research. The compound is supplied as a white to off-white solid with high reported purity and is used as a pharmacological tool to study lysosomal calcium signaling, autophagy, and related cellular pathways.
  • High purity suitable for biochemical and cellular assays.
  • Solid form for convenient handling and storage.
  • Available in small-scale quantities for screening and optimization.
  • Applicable for studies of lysosomal function and autophagy-related mechanisms.
  • Molecular formula C23H31N3O3S and molecular weight 429.58 g·mol⁻¹ for reference.
Catalog No. 50-002-22186 Supplier Medchemexpress LLC Supplier No. HY139426A5MG
(rel)-ML-SI3 is a small-molecule TRPML1/3 inhibitor intended for preclinical and in vitro research. The compound is supplied as a white to off-white solid with high reported purity and is used as a pharmacological tool to study lysosomal calcium signaling, autophagy, and related cellular pathways.
  • High purity suitable for biochemical and cellular assays.
  • Solid form for convenient handling and storage.
  • Available in small-scale quantities for screening and optimization.
  • Applicable for studies of lysosomal function and autophagy-related mechanisms.
  • Molecular formula C23H31N3O3S and molecular weight 429.58 g·mol⁻¹ for reference.