Learn More
Medchemexpress LLC (rel)-Nutlin carboxylic acid | 2760669-71-2 | 10 MG
SDP
Catalog No. 5000341148
Encompass_Preferred
(rel)-Nutlin carboxylic acid is a Nutlin 3-based MDM2 ligand. It is designed to be connected to a protein ligand via a linker, enabling the formation of PROTAC molecules.
  • Functions as a Nutlin 3-based MDM2 ligand
  • Connects to protein ligands via a linker
  • Enables the formation of PROTAC
Catalog No. 50-003-41148 Supplier Medchemexpress LLC Supplier No. HY12883710MG
(rel)-Nutlin carboxylic acid is a Nutlin 3-based MDM2 ligand. It is designed to be connected to a protein ligand via a linker, enabling the formation of PROTAC molecules.
  • Functions as a Nutlin 3-based MDM2 ligand
  • Connects to protein ligands via a linker
  • Enables the formation of PROTAC