Learn More
Medchemexpress LLC (rel)-Nutlin carboxylic acid | 2760669-71-2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12883750MG
Encompass_Preferred
(rel)-Nutlin carboxylic acid is a Nutlin 3-based MDM2 ligand that can be connected to a ligand for protein by a linker to form PROTAC. This product has a molecular formula of C32H32Cl2N4O6 and a molecular weight of 639.53. It is intended for research use only.
  • Nutlin 3-based MDM2 ligand
  • Can be connected to a ligand for protein by a linker to form PROTAC
  • Targets MDM2
  • Targets Ligands for E3 Ligase
Catalog No. 50-003-38213