Learn More
Medchemexpress LLC Rel-paroxetine-d4 hydrochloride | 1217683-35-6 | C19H17D4ClFNO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY151216S1MG
Encompass_Preferred
Rel-Paroxetine-d4 hydrochloride is an isotope-labeled rel-Paroxetine hydrochloride, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
  • Utilized as a tracer for quantitation in drug development processes
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes, potentially influencing pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-75913