Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (rel)-PROTAC ERRα Degrader-1 | 98.2% | 1 MG
SDP

Catalog No. 5000339882
Encompass_Preferred

(rel)-PROTAC ERRα Degrader-1 is a relative configuration of PROTAC ERRα Degrader-1, comprising an MDM2 ligand binding group, a linker, and an estrogen-related receptor alpha (ERRα) binding group. It functions as an estrogen-related receptor alpha (ERRα) degrader.

  • Molecular weight: 1108.92
  • Molecular formula: C₅₄H₄₉Cl₂F₆N₇O₈
  • Appearance: Off-white to light yellow solid
  • Purity: 98.18%
  • Soluble in DMSO at 100 mg/mL
  • Target: Estrogen receptor/ERR; PROTACs
  • Pathway: Vitamin D related/nuclear receptor; PROTAC

Catalog No. 50-003-39882 Supplier Medchemexpress LLC Supplier No. HY128838A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(rel)-PROTAC ERRα Degrader-1 is a relative configuration of PROTAC ERRα Degrader-1, comprising an MDM2 ligand binding group, a linker, and an estrogen-related receptor alpha (ERRα) binding group. It functions as an estrogen-related receptor alpha (ERRα) degrader.

  • Molecular weight: 1108.92
  • Molecular formula: C₅₄H₄₉Cl₂F₆N₇O₈
  • Appearance: Off-white to light yellow solid
  • Purity: 98.18%
  • Soluble in DMSO at 100 mg/mL
  • Target: Estrogen receptor/ERR; PROTACs
  • Pathway: Vitamin D related/nuclear receptor; PROTAC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.