Learn More
Medchemexpress LLC Rel-PROTAC PARP1 degrader | 99.0% | 1145.09 | 10 MG
SDP
Catalog No. 5000436839
Encompass_Preferred
rel-PROTAC PARP1 degrader is the relative configuration of ROTAC PARP1 degrader (HY-114324). ROTAC PARP1 degrader is a PARP1 degrader based on MDM2 E3 ligand.
  • Induces significant PARP1 cleavage.
  • Induces programmed cell death.
  • Inhibits MDA-MB-231 cell line with an IC50 of 6.12 μM at 10 μM within 24 hours.
  • For research use only.
Catalog No. 50-004-36839 Supplier Medchemexpress LLC Supplier No. HY114324A10MG
rel-PROTAC PARP1 degrader is the relative configuration of ROTAC PARP1 degrader (HY-114324). ROTAC PARP1 degrader is a PARP1 degrader based on MDM2 E3 ligand.
  • Induces significant PARP1 cleavage.
  • Induces programmed cell death.
  • Inhibits MDA-MB-231 cell line with an IC50 of 6.12 μM at 10 μM within 24 hours.
  • For research use only.