Learn More
Medchemexpress LLC Rel-Ritonavir-d6 | 1217720-20-1 | 98.7% | C37H42D6N6O5S2 | 500 UG
SDP
Catalog No. 5000432375
Encompass_Preferred
rel-Ritonavir-d6 is the deuterium labeled rel-Ritonavir, a stable isotope of Ritonavir. Ritonavir is an inhibitor of HIV protease, commonly used in the study of HIV infection and AIDS, and also functions as a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM. The incorporation of stable heavy isotopes into drug molecules, such as deuterium in rel-Ritonavir-d6, is largely for quantitation during the drug development process and has shown potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled rel-Ritonavir
  • Inhibitor of HIV protease
  • Used to study HIV infection and AIDS
  • SARS-CoV 3CLpro inhibitor
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32375 Supplier Medchemexpress LLC Supplier No. HY90001S3500UG
rel-Ritonavir-d6 is the deuterium labeled rel-Ritonavir, a stable isotope of Ritonavir. Ritonavir is an inhibitor of HIV protease, commonly used in the study of HIV infection and AIDS, and also functions as a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM. The incorporation of stable heavy isotopes into drug molecules, such as deuterium in rel-Ritonavir-d6, is largely for quantitation during the drug development process and has shown potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled rel-Ritonavir
  • Inhibitor of HIV protease
  • Used to study HIV infection and AIDS
  • SARS-CoV 3CLpro inhibitor
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS