Learn More
Medchemexpress LLC (rel)-Tivantinib | 905853-99-8 | 96.5% | C23H19N3O2 | 1 ML
SDP
Catalog No. 5000433543
Encompass_Preferred
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are significant in the cellular mechanism of non-small cell lung cancer (NSCLC). This product is intended for research use only.
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β
  • Important in the cellular mechanism of non-small cell lung cancer (NSCLC)
  • For research use only
Catalog No. 50-004-33543 Supplier Medchemexpress LLC Supplier No. HY7749310MM/1ML
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are significant in the cellular mechanism of non-small cell lung cancer (NSCLC). This product is intended for research use only.
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β
  • Important in the cellular mechanism of non-small cell lung cancer (NSCLC)
  • For research use only