Learn More
Medchemexpress LLC (rel)-Tivantinib | 905853-99-8 | 96.5% | C23H19N3O2 | 100 MG
SDP
Catalog No. 5000434938
Encompass_Preferred
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β
Catalog No. 50-004-34938 Supplier Medchemexpress LLC Supplier No. HY77493100MG
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective inhibitor of c-MET
  • Targets GSK3α and GSK3β