Learn More
Medchemexpress LLC (rel)-Tivantinib | 905853-99-8 | MFCD11977597 | 96.45% | C23H19N3O2 | 25 MG
SDP
Catalog No. 5000359000
Encompass_Preferred
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective c-MET inhibitor
  • Targets GSK3α and GSK3β
  • Important in non-small cell lung cancer (NSCLC) cellular mechanisms
Catalog No. 50-003-59000 Supplier Medchemexpress LLC Supplier No. 25MGHY77493
(rel)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective c-MET inhibitor
  • Targets GSK3α and GSK3β
  • Important in non-small cell lung cancer (NSCLC) cellular mechanisms