Learn More
Medchemexpress LLC rel-VU6021625 | 2307718-96-1 | 99.67% | 432.56 | 25 MG
SDP
Catalog No. 5000394307
Encompass_Preferred
rel-VU6021625 is the relative configuration of VU6021625. VU6021625 is identified as a potent and selective mAChR M4 antagonist, exhibiting IC50 values of 0.44 nM for human M4 and 57 nM for rat M4. The product is intended for research use only.
  • Potent and selective mAChR M4 antagonist
  • Relative configuration of VU6021625
  • IC50 values: 0.44 nM (human M4), 57 nM (rat M4)
Catalog No. 50-003-94307 Supplier Medchemexpress LLC Supplier No. 25MGHY160440
rel-VU6021625 is the relative configuration of VU6021625. VU6021625 is identified as a potent and selective mAChR M4 antagonist, exhibiting IC50 values of 0.44 nM for human M4 and 57 nM for rat M4. The product is intended for research use only.
  • Potent and selective mAChR M4 antagonist
  • Relative configuration of VU6021625
  • IC50 values: 0.44 nM (human M4), 57 nM (rat M4)