Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rel-VU6021625 | 2307718-96-1 | 99.7% | 432.56 | C25H32N6O | 10 MG
SDP

Catalog No. 5000285483
Encompass_Preferred

rel-VU6021625 is the relative stereochemical isomer of VU6021625, a potent and selective antagonist of the muscarinic acetylcholine receptor M4. It is supplied as a white to off-white solid intended for research use only; reported activity includes IC50 = 0.44 nM (human M4) and 57 nM (rat M4). Chemical identifiers include formula C25H32N6O, molecular weight 432.56, CAS 2307718-96-1, and high purity (~99.7%).

  • Selective mAChR M4 antagonist with nanomolar potency.
  • Intended for research use only.
  • White to off-white solid with high purity (~99.7%).
  • Chemical formula C25H32N6O; molecular weight 432.56.
  • Available in 10 MG quantities.
  • Storage: powder at -20°C for long term; in solvent store at -80°C for extended periods.

Catalog No. 50-002-85483 Supplier Medchemexpress LLC Supplier No. HY16044010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rel-VU6021625 is the relative stereochemical isomer of VU6021625, a potent and selective antagonist of the muscarinic acetylcholine receptor M4. It is supplied as a white to off-white solid intended for research use only; reported activity includes IC50 = 0.44 nM (human M4) and 57 nM (rat M4). Chemical identifiers include formula C25H32N6O, molecular weight 432.56, CAS 2307718-96-1, and high purity (~99.7%).

  • Selective mAChR M4 antagonist with nanomolar potency.
  • Intended for research use only.
  • White to off-white solid with high purity (~99.7%).
  • Chemical formula C25H32N6O; molecular weight 432.56.
  • Available in 10 MG quantities.
  • Storage: powder at -20°C for long term; in solvent store at -80°C for extended periods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.