Learn More
Medchemexpress LLC rel-Zotatifin | 2098191-54-7 | 97.3% | 487.56 g/mol | C28H29N3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY112163A5MG
Encompass_Preferred
rel-Zotatifin is a research-grade small-molecule inhibitor of the eukaryotic initiation factor eIF4A, supplied as the racemic isomer for laboratory studies. It is characterized by a molecular formula of C28H29N3O5, molecular weight of 487.56 g/mol, and high purity suitable for biochemical and cellular assays.
  • Racemic isomer with eIF4A inhibitory activity.
  • High supplied purity for research applications.
  • Supplied as a small, convenient 5 mg package for screening and assay development.
  • Suitable for biochemical and cell-based translation studies.
  • Molecular weight and formula provided for analytical confirmation.
Catalog No. 50-002-63313