Learn More
Aviva Systems Biology Corp RELA Antibody - N-terminal region (P100779_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100779_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RELA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RELA. Alternate names: p65, NFKB3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62710