Learn More
Aviva Systems Biology Corp RELA Antibody - middle region (ARP30371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RELA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RELA. Alternate names: MGC131774, NFKB3, p65ApplicationWB, IHC. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54160