Learn More
Medchemexpress LLC Relamorelin (TFA) | 2863659-22-5 | 99.8% | 905.00 | 100 MG
SDP
Catalog No. 5000401744
Encompass_Preferred
Relamorelin (RM-131) TFA, a pentapeptide ghrelin analog, is a selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist with a Ki of 0.42 nM for the GHS-1a receptor. It is centrally penetrant, increases growth hormone levels, and accelerates gastric emptying. Relamorelin TFA has potential for research into cachexia, gastroparesis, and gastric/intestinal dysmobility disorders.
  • Selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist.
  • Ki of 0.42 nM for GHS-1a receptor.
  • Centrally penetrant.
  • Increases growth hormone levels.
  • Accelerates gastric emptying.
  • Potential for cachexia, gastroparesis, and gastric/intestinal dysmobility disorders research.
  • Purity: 99.81%.
Catalog No. 50-004-01744 Supplier Medchemexpress LLC Supplier No. HY19884B100MG
Relamorelin (RM-131) TFA, a pentapeptide ghrelin analog, is a selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist with a Ki of 0.42 nM for the GHS-1a receptor. It is centrally penetrant, increases growth hormone levels, and accelerates gastric emptying. Relamorelin TFA has potential for research into cachexia, gastroparesis, and gastric/intestinal dysmobility disorders.
  • Selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist.
  • Ki of 0.42 nM for GHS-1a receptor.
  • Centrally penetrant.
  • Increases growth hormone levels.
  • Accelerates gastric emptying.
  • Potential for cachexia, gastroparesis, and gastric/intestinal dysmobility disorders research.
  • Purity: 99.81%.