Learn More
Medchemexpress LLC Relamorelin (TFA) | 2863659-22-5 | 99.8% | 905.00 | 25 MG
SDP
Catalog No. 5000402233
Encompass_Preferred
Relamorelin TFA is a pentapeptide ghrelin analog and a selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist with a Ki of 0.42 nM for the GHS-1a receptor. It is centrally penetrant, increases growth hormone levels, and accelerates gastric emptying. This compound has potential for research into cachexia, gastroparesis, and gastric/intestinal dysmobility disorders.
  • Pentapeptide ghrelin analog
  • Selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist
  • Ki of 0.42 nM for GHS-1a receptor
  • Centrally penetrant
  • Increases growth hormone levels
  • Accelerates gastric emptying
  • Shows ~3 times greater affinity for GHS-1a (Ki=0.42 nM) than native ghrelin (Ki=1.12 nM)
  • 6 times more potent (EC50=0.71 nM) in activating the GHS-1a receptor than native ghrelin (EC50=4.2 nM) in vitro by calcium mobilization
  • Solid appearance, white to off-white color
  • For research use only.
Catalog No. 50-004-02233 Supplier Medchemexpress LLC Supplier No. 25MGHY19884B
Relamorelin TFA is a pentapeptide ghrelin analog and a selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist with a Ki of 0.42 nM for the GHS-1a receptor. It is centrally penetrant, increases growth hormone levels, and accelerates gastric emptying. This compound has potential for research into cachexia, gastroparesis, and gastric/intestinal dysmobility disorders.
  • Pentapeptide ghrelin analog
  • Selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist
  • Ki of 0.42 nM for GHS-1a receptor
  • Centrally penetrant
  • Increases growth hormone levels
  • Accelerates gastric emptying
  • Shows ~3 times greater affinity for GHS-1a (Ki=0.42 nM) than native ghrelin (Ki=1.12 nM)
  • 6 times more potent (EC50=0.71 nM) in activating the GHS-1a receptor than native ghrelin (EC50=4.2 nM) in vitro by calcium mobilization
  • Solid appearance, white to off-white color
  • For research use only.