Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Relamorelin TFA | 2863659-22-5 | 99.8% | 905.00 | C45H51F3N8O7S | 10 MG
SDP

Catalog No. 5000297326
Encompass_Preferred

Relamorelin TFA is the trifluoroacetate salt of Relamorelin (RM-131), a pentapeptide ghrelin/growth hormone secretagogue receptor agonist supplied as a solid for laboratory research. It stimulates growth hormone release and accelerates gastric emptying, and is used in preclinical investigations of gastrointestinal motility and cachexia.

  • High purity of 99.8%.
  • Molecular weight of 905.00 g/mol.
  • CAS number 2863659-22-5.
  • Supplied as a solid suitable for bench-top handling and formulation.
  • Selective GHSR agonist that increases growth hormone release and accelerates gastric emptying.
  • Intended for preclinical research into gastroparesis and cachexia models.

Catalog No. 50-002-97326 Supplier Medchemexpress LLC Supplier No. HY19884B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Relamorelin TFA is the trifluoroacetate salt of Relamorelin (RM-131), a pentapeptide ghrelin/growth hormone secretagogue receptor agonist supplied as a solid for laboratory research. It stimulates growth hormone release and accelerates gastric emptying, and is used in preclinical investigations of gastrointestinal motility and cachexia.

  • High purity of 99.8%.
  • Molecular weight of 905.00 g/mol.
  • CAS number 2863659-22-5.
  • Supplied as a solid suitable for bench-top handling and formulation.
  • Selective GHSR agonist that increases growth hormone release and accelerates gastric emptying.
  • Intended for preclinical research into gastroparesis and cachexia models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.