Learn More
Thermo Scientific™ Relative Humidity System Option
GSA_VA
Catalog No. 13762730 Shop All Thermo Scientific Products
Change view
Click to view available options
Installation:
Factory Installed
Catalog No. Installation
13-762-730 Factory Installed
1 options
Catalog No. 13-762-730 Supplier Thermo Scientific™ Supplier No. 1900139
For Use With: Environmental Chambers (3948 and 3907)
Use Thermo Scientific™ Relative Humidity System Options for Environmental Chambers to address specific requirements to the relative humidity system.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Heatless Dryer Kit for sub-ambient chamber RH
For Use With (Equipment) Environmental Chambers 3948/3907
Lighting LED
Type Relative Humidity System
Installation Factory Installed