Learn More
Bachem Relaxin H3 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4039569.1000
Encompass_Preferred
Relaxin H3 (human) Trifluoroacetate, (H-Asp-Val-Leu-Ala-Gly-Leu-Ser-Ser-Ser-Cys-Cys-Lys-Trp-Gly-Cys-Ser-Lys-Ser-Glu-Ile-Ser-Ser-Leu-Cys-OH)A(H-Arg-Ala-Ala-Pro-Tyr-Gly-Val-Arg-Leu-Cys-Gly-Arg-Glu-Phe-Ile-Arg-Ala-Val-Ile-Phe-Thr-Cys-Gly-Gly-Ser-Arg-Trp-OH)B, 1158181-62-4, C237H374N70O69S6, Mr 5500.40, 1mg. Relaxin H3 is a specific ligand for the leucine-rich repeat-containing G protein-coupled receptor LGR7. Hossain et al. demonstrated that relaxin H3 exerts antifibrotic actions via the relaxin receptor RXFP1 and may enhance the collagen-inhibitory effects of relaxin H2.
Catalog No. 50-194-6562