Learn More
Grainger Relay 24VDC
Catalog No. 502766687
Encompass_Preferred
The Siemens IEC Control Relay is a 24V DC relay with 13A contact rating. It has 2 NO/NC contacts and a built-in LED indicator. The relay has screw terminal connections and can switch both AC and DC loads up to 480V AC. It is 35mm wide and mounts on a standard DIN rail. The relay has a transparent dust cover and plug-in socket. It is rated for 10 million mechanical operations. The operating temperature range is -25 to 60°C.
Catalog No. 50-276-6687 Supplier Grainger Supplier No. 16X421
The Siemens IEC Control Relay is a 24V DC relay with 13A contact rating. It has 2 NO/NC contacts and a built-in LED indicator. The relay has screw terminal connections and can switch both AC and DC loads up to 480V AC. It is 35mm wide and mounts on a standard DIN rail. The relay has a transparent dust cover and plug-in socket. It is rated for 10 million mechanical operations. The operating temperature range is -25 to 60°C.