Learn More
Grainger Relay DPDT
Catalog No. 502766101
Encompass_Preferred
The Edwards Signaling 16X423 is a DPDT relay that has a coil voltage of 24VDC and contact ratings of 10A at 250VAC and 28VDC. This relay has two sets of Form C contacts, meaning there are both normally open and normally closed contacts. The relay has an LED that illuminates when the coil is energized. It can switch up to 10 amps resistive loads. This relay has a plug-in mounting style and quick connect or solder terminals. It is well suited for HVAC, security, fire alarm, access control, pump control and other industrial control applications.
Catalog No. 50-276-6101 Supplier Grainger Supplier No. 16X423
The Edwards Signaling 16X423 is a DPDT relay that has a coil voltage of 24VDC and contact ratings of 10A at 250VAC and 28VDC. This relay has two sets of Form C contacts, meaning there are both normally open and normally closed contacts. The relay has an LED that illuminates when the coil is energized. It can switch up to 10 amps resistive loads. This relay has a plug-in mounting style and quick connect or solder terminals. It is well suited for HVAC, security, fire alarm, access control, pump control and other industrial control applications.