Learn More
Grainger Relay SPDT
Catalog No. 502767543
Encompass_Preferred
The Edwards Signaling 115V AC/230V AC/24V DC Relay is a general purpose relay that can be used for a variety of signaling and control applications. It has a 10 amp contact rating and is offered in 1, 2, 3, or 4 pole configurations. The relay has quick connect terminals for easy wiring and LED indication shows when the relay is energized. It can switch both AC and DC loads up to its 10A rating. The relay mounts via 35mm DIN rail or can be screw mounted. It meets UL and CSA standards. With its versatile operating voltage and multiple contact configurations, this relay provides flexible control options for industrial, commercial, and OEM applications.
Catalog No. 50-276-7543 Supplier Grainger Supplier No. 16X420
The Edwards Signaling 115V AC/230V AC/24V DC Relay is a general purpose relay that can be used for a variety of signaling and control applications. It has a 10 amp contact rating and is offered in 1, 2, 3, or 4 pole configurations. The relay has quick connect terminals for easy wiring and LED indication shows when the relay is energized. It can switch both AC and DC loads up to its 10A rating. The relay mounts via 35mm DIN rail or can be screw mounted. It meets UL and CSA standards. With its versatile operating voltage and multiple contact configurations, this relay provides flexible control options for industrial, commercial, and OEM applications.