Learn More
Grainger Relay SPDT
Catalog No. 502765186
Encompass_Preferred
The Edwards Signaling 115V AC/24V AC/24V DC Relay is a general purpose relay that can switch 115V AC, 24V AC, or 24V DC loads. It has a 10 amp contact rating and plug-in socket terminals for easy installation. The relay has an enclosed coil and clear dust cover for protection. It can be mounted on a flat surface or snapped onto 35mm DIN rail. This industrial-grade electromechanical relay is well-suited for controlling motors, lights, pumps, valves, and other equipment in commercial buildings and industrial facilities.
Catalog No. 50-276-5186 Supplier Grainger Supplier No. 16X424
The Edwards Signaling 115V AC/24V AC/24V DC Relay is a general purpose relay that can switch 115V AC, 24V AC, or 24V DC loads. It has a 10 amp contact rating and plug-in socket terminals for easy installation. The relay has an enclosed coil and clear dust cover for protection. It can be mounted on a flat surface or snapped onto 35mm DIN rail. This industrial-grade electromechanical relay is well-suited for controlling motors, lights, pumps, valves, and other equipment in commercial buildings and industrial facilities.