Learn More
Aviva Systems Biology Corp RELB Antibody - middle region (ARP36969_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36969_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RELB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse RELB. Alternate names: shepApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56299