Learn More
Cayman Chemical ReldesemtIv 10mg
SDP
Catalog No. 502943637
Encompass_Preferred
An activator of fast skeletal troponin; increases ATPase activity in isolated rabbit psoas muscle myofibril preparations (AC40 = 0.6 µM); selective for fast skeletal troponin over cardiac and slow skeletal troponin; selective for fast skeletal troponin over a panel of 79 enzymes, receptors, ion channels, and transporters at 100 µM; increases muscle tension in a skinned fast skeletal muscle fiber assay (DF30 = 0.8 µM); increases running time in the rotarod test, indicating increased exercise performance, in a rat model of heart failure-induced skeletal myopathy at 10 mg/kg, p.o.
Catalog No. 50-294-3637 Supplier Cayman Chemical Supplier No. 4184010 MG
An activator of fast skeletal troponin; increases ATPase activity in isolated rabbit psoas muscle myofibril preparations (AC40 = 0.6 µM); selective for fast skeletal troponin over cardiac and slow skeletal troponin; selective for fast skeletal troponin over a panel of 79 enzymes, receptors, ion channels, and transporters at 100 µM; increases muscle tension in a skinned fast skeletal muscle fiber assay (DF30 = 0.8 µM); increases running time in the rotarod test, indicating increased exercise performance, in a rat model of heart failure-induced skeletal myopathy at 10 mg/kg, p.o.