Learn More
TARGETMOL CHEMICALS INC RELDESEMTIV 5MG
SDP
Catalog No. 502581160
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reldesemtiv (CK-2127107) is a selective activator of fast skeletal muscle troponin with an EC50 of 3.4 uM. Reldesemtiv increases exercise performance in the heart failure model. purity: 95%
Catalog No. 50-258-1160 Supplier TARGETMOL CHEMICALS INC Supplier No. T167285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reldesemtiv (CK-2127107) is a selective activator of fast skeletal muscle troponin with an EC50 of 3.4 uM. Reldesemtiv increases exercise performance in the heart failure model. purity: 95%