Learn More
MESA LABS BOZEMAN MFG FACILITY RELEASAT TBES W/STPS GAS 100PK
SDP
Supplier:  MESA LABS BOZEMAN MFG FACILITY RG/100
Encompass
Releasat Rapid-Readout Color Change EtO Culturing Kit, Incubation: 72 Hours at 37+/-1 deg.C, Population: 106, 100/BX
Catalog No. NC9249977